Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00005846.mm.1 |
NM_001034907.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005846.mm.1 |
NM_001034907.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005846.mm.1 |
TC0X00000912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005847.mm.1 |
NM_001034907.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005847.mm.1 |
NM_001034907.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005847.mm.1 |
TC0X00000912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005851.mm.1 |
NM_001034907.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005851.mm.1 |
NM_001034907.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005851.mm.1 |
TC0X00000912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005852.mm.1 |
NM_001034907.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005852.mm.1 |
NM_001034907.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005852.mm.1 |
TC0X00000912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005854.mm.1 |
NM_001034907.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005854.mm.1 |
NM_001034907.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005854.mm.1 |
TC0X00000912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005845.mm.1 |
NM_001034907.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005845.mm.1 |
NM_001034907.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005845.mm.1 |
TC0X00000912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005843.mm.1 |
NM_001034907.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005843.mm.1 |
NM_001034907.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005843.mm.1 |
TC0X00000912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005844.mm.1 |
NM_001034907.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005844.mm.1 |
NM_001034907.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005844.mm.1 |
TC0X00000912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005849.mm.1 |
NM_001034907.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005849.mm.1 |
NM_001034907.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005849.mm.1 |
TC0X00000912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005853.mm.1 |
NM_001034907.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005853.mm.1 |
NM_001034907.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005853.mm.1 |
TC0X00000912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005855.mm.1 |
NM_001034907.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005855.mm.1 |
NM_001034907.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005855.mm.1 |
TC0X00000912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005848.mm.1 |
NM_001034907.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005848.mm.1 |
NM_001034907.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005848.mm.1 |
TC0X00000912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005857.mm.1 |
NM_001034907.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005857.mm.1 |
NM_001034907.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005857.mm.1 |
TC0X00000912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005856.mm.1 |
NM_001034907.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005856.mm.1 |
NM_001034907.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005856.mm.1 |
TC0X00000912.mm.1 |
MTA TC |