Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad54l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030730.2 | NP_109655.2 | 6 - 7 | 1108 | 155 | |
GenBank mRNA | AJ132389.1 | - | 6 - 7 | 1009 | 155 | |
AK083903.1 | - | 6 - 7 | 1089 | 155 | ||
AK143506.1 | - | 5 - 6 | 724 | 155 | ||
AK155151.1 | - | 6 - 7 | 1065 | 155 | ||
BC133714.1 | - | 6 - 7 | 957 | 40143565 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |