Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700011794.mm.1 |
NM_011104.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011794.mm.1 |
NM_011104.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011794.mm.1 |
TC1700001289.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011793.mm.1 |
NM_011104.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011793.mm.1 |
NM_011104.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011793.mm.1 |
TC1700001289.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011795.mm.1 |
NM_011104.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011795.mm.1 |
NM_011104.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011795.mm.1 |
TC1700001289.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011801.mm.1 |
NM_011104.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011801.mm.1 |
NM_011104.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011801.mm.1 |
TC1700001289.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011791.mm.1 |
NM_011104.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011791.mm.1 |
NM_011104.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011791.mm.1 |
TC1700001289.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011790.mm.1 |
NM_011104.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011790.mm.1 |
NM_011104.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011790.mm.1 |
TC1700001289.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700011792.mm.1 |
NM_011104.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011792.mm.1 |
NM_011104.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700011792.mm.1 |
TC1700001289.mm.1 |
MTA TC |