Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253831.1 | NP_001240760.1 | 14 - 15 | 1427 | 111 | |
NM_001253834.1 | NP_001240763.1 | 15 - 16 | 1518 | 111 | ||
NM_001253836.1 | NP_001240765.1 | 14 - 15 | 1450 | 111 | ||
NM_175025.4 | NP_778190.3 | 15 - 16 | 1633 | 111 | ||
GenBank mRNA | AJ551270.1 | - | 13 - 14 | 1215 | 111 | |
AK028393.1 | - | 14 - 15 | 1396 | 111 | ||
AK133703.1 | - | 14 - 15 | 1428 | 111 | ||
AK159918.1 | - | 15 - 16 | 1584 | 111 | ||
AK163049.1 | - | 15 - 16 | 1479 | 111 | ||
AK173154.1 | - | 13 - 14 | 1206 | 111 | ||
BC043091.1 | - | 15 - 16 | 1599 | 6405251 | 111 | |
BC043706.1 | - | 15 - 16 | 1579 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |