Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081309.1 | NP_001074778.1 | 7 - 8 | 2421 | 77 | |
GenBank mRNA | AK050333.1 | - | 7 - 8 | 2391 | 77 | |
AK054559.1 | - | 7 - 8 | 2438 | 77 | ||
AK087938.1 | - | 7 - 8 | 2412 | 77 | ||
AK169508.1 | - | 6 - 7 | 3208 | 77 | ||
BC157947.1 | - | 7 - 8 | 2246 | 9007281 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |