Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033444.2 | NP_001028616.1 | 1 - 2 | 121 | 87 | |
GenBank mRNA | AK136569.1 | - | 1 - 2 | 122 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Protease Mus musculus Probe spans exons |