Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900010050.mm.1 |
NM_001042607.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010050.mm.1 |
NM_001042607.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010050.mm.1 |
NM_001284258.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010050.mm.1 |
NM_013649.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010050.mm.1 |
NM_013649.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010050.mm.1 |
TC0900001295.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900010051.mm.1 |
NM_001042607.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010051.mm.1 |
NM_001042607.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010051.mm.1 |
NM_001284258.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010051.mm.1 |
NM_013649.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010051.mm.1 |
NM_013649.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010051.mm.1 |
TC0900001295.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900010054.mm.1 |
NM_001042607.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010054.mm.1 |
NM_001042607.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010054.mm.1 |
NM_013649.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010054.mm.1 |
NM_013649.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010054.mm.1 |
TC0900001295.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900010048.mm.1 |
NM_001042607.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010048.mm.1 |
NM_001042607.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010048.mm.1 |
NM_001284258.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010048.mm.1 |
NM_013649.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010048.mm.1 |
NM_013649.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900010048.mm.1 |
TC0900001295.mm.1 |
MTA TC |