Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021287.1 | NP_067262.1 | 34 - 35 | 7163 | 74 | |
GenBank mRNA | AK147671.1 | - | 34 - 35 | 7179 | 74 | |
AK172931.1 | - | 24 - 25 | 5472 | 74 | ||
BC048851.1 | - | 12 - 13 | 1981 | 74 | ||
BC079628.1 | - | 12 - 13 | 2115 | 74 | ||
BC079860.1 | - | 34 - 35 | 7163 | 6834745 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |