Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162870.1 | NP_001156342.1 | 14 - 15 | 1658 | 78 | |
NM_016962.2 | NP_058658.2 | 14 - 15 | 1655 | 78 | ||
GenBank mRNA | AJ246002.1 | - | 14 - 15 | 1280 | 78 | |
AK007793.1 | - | 14 - 15 | 1435 | 78 | ||
AK129282.1 | - | 14 - 15 | 1632 | 78 | ||
BC046286.1 | - | 14 - 15 | 1655 | 6441742 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |