Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162870.1 | NP_001156342.1 | 8 - 9 | 1213 | 126 | |
NM_016962.2 | NP_058658.2 | 8 - 9 | 1210 | 126 | ||
GenBank mRNA | AJ246002.1 | - | 8 - 9 | 835 | 126 | |
AK007793.1 | - | 8 - 9 | 990 | 126 | ||
AK044900.1 | - | 8 - 9 | 1132 | 126 | ||
AK129282.1 | - | 8 - 9 | 1187 | 126 | ||
BC046286.1 | - | 8 - 9 | 1210 | 6441742 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |