Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015827.2 | NP_056642.1 | 14 - 15 | 1769 | 131 | |
GenBank mRNA | AF043120.1 | - | 3 - 4 | 314 | 131 | |
AK088064.1 | - | 14 - 15 | 1770 | 131 | ||
AK153352.1 | - | 14 - 15 | 1770 | 131 | ||
BC006675.1 | - | 14 - 15 | 1756 | 3499585 | 131 | |
BC010842.1 | - | 14 - 15 | 1758 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |