Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cspg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166273.1 | NP_001159745.1 | 3 - 4 | 3195 | 84 | |
NM_013884.3 | NP_038912.3 | 3 - 4 | 3195 | 84 | ||
GenBank mRNA | AF133700.1 | - | 3 - 4 | 1380 | 84 | |
AF461090.1 | - | 3 - 4 | 3189 | 84 | ||
AF461091.1 | - | 2 - 3 | 1513 | 84 | ||
AF461092.1 | - | 3 - 4 | 3189 | 84 | ||
AK053891.1 | - | 2 - 3 | 777 | 84 | ||
BC055736.1 | - | 3 - 4 | 1527 | 6412504 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |