Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013599.3 | NP_038627.1 | 5 - 6 | 857 | 83 | |
GenBank mRNA | AK004651.1 | - | 5 - 6 | 838 | 83 | |
AK142787.1 | - | 5 - 6 | 839 | 83 | ||
AK159292.1 | - | 5 - 6 | 839 | 83 | ||
AK161176.1 | - | 5 - 6 | 838 | 83 | ||
BC046991.1 | - | 5 - 6 | 825 | 6309245 | 83 | |
D12712.1 | - | 5 - 6 | 846 | 83 | ||
S67830.1 | - | 5 - 6 | 866 | 83 | ||
X72795.1 | - | 3 - 4 | 424 | 83 | ||
Z27231.1 | - | 5 - 6 | 838 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |