Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167746.1 | NP_001161218.1 | 44 - 45 | 6770 | 68 | |
GenBank mRNA | AK173343.1 | - | 2 - 3 | 197 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |