Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128086.2 | NP_001121558.2 | 21 - 22 | 3415 | 79 | |
NM_001177396.1 | NP_001170867.1 | 8 - 9 | 1318 | 79 | ||
NM_001177397.1 | NP_001170868.1 | 7 - 8 | 861 | 79 | ||
NM_001177398.1 | NP_001170869.1 | 7 - 8 | 1141 | 79 | ||
NM_181398.3 | NP_852063.1 | 6 - 7 | 1066 | 79 | ||
GenBank mRNA | AK032061.1 | - | 5 - 6 | 517 | 79 | |
AK032211.1 | - | 7 - 8 | 1143 | 79 | ||
AK034154.1 | - | 8 - 9 | 881 | 79 | ||
AK038609.1 | - | 5 - 6 | 745 | 79 | ||
AK049899.1 | - | 6 - 7 | 1011 | 79 | ||
AK132228.1 | - | 8 - 9 | 1285 | 79 | ||
AK163750.1 | - | 6 - 7 | 1040 | 79 | ||
BC098373.1 | - | 7 - 8 | 861 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |