Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mvd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138656.2 | NP_619597.2 | 1 - 2 | 131 | 58 | |
NR_028354.1 | 1 - 2 | 131 | 58 | |||
GenBank mRNA | AJ309922.1 | - | 1 - 2 | 88 | 58 | |
AK089354.1 | - | 1 - 2 | 133 | 58 | ||
AK134803.1 | - | 1 - 2 | 120 | 58 | ||
AK153439.1 | - | 1 - 2 | 106 | 58 | ||
AK154883.1 | - | 1 - 2 | 133 | 58 | ||
AK170007.1 | - | 1 - 2 | 68 | 58 | ||
AK170773.1 | - | 1 - 2 | 127 | 58 | ||
BC008526.1 | - | 1 - 2 | 99 | 3495424 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |