Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033321.3 | NP_201578.2 | 9 - 10 | 1046 | 87 | |
GenBank mRNA | AK134662.1 | - | 9 - 10 | 1006 | 87 | |
BC138189.1 | - | 9 - 10 | 1046 | 8861208 | 87 | |
BC145181.1 | - | 9 - 10 | 1046 | 9053732 | 87 | |
JX570808.1 | - | 8 - 9 | 1309 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |