Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpgrip1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173431.2 | NP_775607.2 | 23 - 24 | 3491 | 72 | |
GenBank mRNA | AJ344253.1 | - | 23 - 24 | 3531 | 72 | |
AK029911.1 | - | 23 - 24 | 3496 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |