Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afg3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054070.3 | NP_473411.2 | 11 - 12 | 1445 | 70 | |
NR_028260.1 | 11 - 12 | 1445 | 70 | |||
GenBank mRNA | AF329695.1 | - | 11 - 12 | 1386 | 70 | |
AK159647.1 | - | 11 - 12 | 1409 | 70 | ||
AK167964.1 | - | 11 - 12 | 1420 | 70 | ||
AK168244.1 | - | 11 - 12 | 1419 | 70 | ||
BC043320.1 | - | 2 - 3 | 267 | 70 | ||
BC056978.1 | - | 11 - 12 | 1404 | 6821746 | 70 | |
BC060064.1 | - | 11 - 12 | 1404 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |