Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009974.3 | NP_034104.1 | 8 - 9 | 1104 | 123 | |
GenBank mRNA | AF012251.1 | - | 8 - 9 | 829 | 123 | |
AJ001420.1 | - | 8 - 9 | 1074 | 123 | ||
AK002927.1 | - | 8 - 9 | 1112 | 123 | ||
AK030035.1 | - | 2 - 3 | 144 | 123 | ||
AK076042.1 | - | 8 - 9 | 1065 | 123 | ||
BC047143.1 | - | 8 - 9 | 1094 | 123 | ||
BC057862.1 | - | 8 - 9 | 764 | 5316132 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |