Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024196.3 | NP_077158.1 | 4 - 5 | 621 | 74 | |
GenBank mRNA | AK006877.1 | - | 4 - 5 | 618 | 74 | |
AK036751.1 | - | 533 | 74 | |||
AK076124.1 | - | 4 - 5 | 622 | 74 | ||
AK085702.1 | - | 619 | 74 | |||
AK158274.1 | - | 618 | 74 | |||
AK168985.1 | - | 622 | 74 | |||
AK172349.1 | - | 618 | 74 | |||
AK172372.1 | - | 50 | 74 | |||
BC034504.1 | - | 603 | 4192736 | 74 | ||
BC040089.1 | - | 4 - 5 | 584 | 3991310 | 74 | |
BC086617.1 | - | 607 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |