Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164499.1 | NP_001157971.1 | 6 - 7 | 1077 | 152 | |
GenBank mRNA | BC144797.1 | - | 6 - 7 | 1035 | 9053345 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |