Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145857.2 | NP_665856.2 | 9 - 10 | 2875 | 65 | |
GenBank mRNA | AF520774.1 | - | 9 - 10 | 2920 | 65 | |
AY160220.1 | - | 8 - 9 | 2804 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mus musculus Probe spans exons |