Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128170.2 | NP_001121642.1 | 10 - 11 | 2045 | 80 | |
NM_001128171.2 | NP_001121643.1 | 10 - 11 | 2170 | 80 | ||
NM_001276279.1 | NP_001263208.1 | 8 - 9 | 1621 | 80 | ||
NM_173369.3 | NP_775545.1 | 9 - 10 | 2036 | 80 | ||
GenBank mRNA | AK039054.1 | - | 11 - 12 | 2174 | 80 | |
AK048183.1 | - | 3 - 4 | 590 | 80 | ||
AK122389.1 | - | 10 - 11 | 2151 | 80 | ||
AK133479.1 | - | 9 - 10 | 2000 | 80 | ||
AK147204.1 | - | 10 - 11 | 2014 | 80 | ||
AK147426.1 | - | 10 - 11 | 2009 | 80 | ||
BC028885.1 | - | 2 - 3 | 1691 | 80 | ||
BC042438.1 | - | 9 - 10 | 1989 | 3983771 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |