Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037665.2 | NP_001032754.1 | 2 - 3 | 228 | 70 | |
GenBank mRNA | AK147054.1 | - | 2 - 3 | 166 | 70 | |
BC025141.1 | - | 2 - 3 | 191 | 70 | ||
BC055796.1 | - | 2 - 3 | 137 | 6412519 | 70 | |
BC062970.1 | - | 2 - 3 | 137 | 6412519 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |