Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028805.2 | NP_083081.2 | 6 - 7 | 795 | 69 | |
GenBank mRNA | AK010364.1 | - | 6 - 7 | 726 | 69 | |
AK031630.1 | - | 6 - 7 | 707 | 69 | ||
AK034103.1 | - | 6 - 7 | 715 | 69 | ||
AK037858.1 | - | 617 | 69 | |||
AK049345.1 | - | 6 - 7 | 709 | 69 | ||
AK087973.1 | - | 6 - 7 | 708 | 69 | ||
AK136322.1 | - | 6 - 7 | 715 | 69 | ||
BC045200.1 | - | 5 - 6 | 706 | 5322489 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |