Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145831.1 | NP_001139303.1 | 14 - 15 | 1904 | 65 | |
NM_001145832.1 | NP_001139304.1 | 10 - 11 | 1418 | 65 | ||
NM_010631.2 | NP_034761.3 | 12 - 13 | 1761 | 65 | ||
GenBank mRNA | AB001457.1 | - | 1 - 2 | 50 | 65 | |
AF013118.2 | - | 8 - 9 | 1397 | 65 | ||
AK132895.1 | - | 10 - 11 | 1418 | 65 | ||
AK135926.1 | - | 7 - 8 | 960 | 65 | ||
AK166482.1 | - | 14 - 15 | 1884 | 65 | ||
BC004069.1 | - | 4 - 5 | 509 | 65 | ||
BC016118.1 | - | 7 - 8 | 965 | 65 | ||
BC023374.1 | - | 5 - 6 | 553 | 65 | ||
BC070429.1 | - | 12 - 13 | 1761 | 6810893 | 65 | |
D49546.1 | - | 12 - 13 | 1776 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |