Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hexdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001333.2 | NP_001001333.2 | 6 - 7 | 1100 | 59 | |
NM_001146073.1 | NP_001139545.1 | 5 - 6 | 686 | 59 | ||
GenBank mRNA | AK154237.1 | - | 6 - 7 | 1074 | 59 | |
BC069960.1 | - | 5 - 6 | 638 | 5007884 | 59 | |
FM204885.1 | - | 5 - 6 | 637 | 59 | ||
FM204886.1 | - | 5 - 6 | 637 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |