Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114347.1 | NP_001107819.1 | 5 - 6 | 756 | 102 | |
NM_001286599.1 | NP_001273528.1 | 5 - 6 | 756 | 102 | ||
GenBank mRNA | AJ276961.1 | - | 5 - 6 | 756 | 102 | |
AK034560.1 | - | 5 - 6 | 719 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |