Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900012584.mm.1 |
NM_001081960.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012584.mm.1 |
NM_001081960.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012584.mm.1 |
NM_001114347.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012584.mm.1 |
NM_001114347.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012584.mm.1 |
NM_001286599.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012584.mm.1 |
NM_001286600.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012584.mm.1 |
NM_001286601.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012584.mm.1 |
NM_001286602.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012584.mm.1 |
NM_001286603.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012584.mm.1 |
NM_029633.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012584.mm.1 |
NM_029633.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012584.mm.1 |
TC0900001504.mm.1 |
MTA TC |