Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100028158.mm.1 |
NM_198862.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028158.mm.1 |
NM_198862.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028158.mm.1 |
TC1100003069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028157.mm.1 |
NM_198862.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028157.mm.1 |
NM_198862.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028157.mm.1 |
TC1100003069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028160.mm.1 |
NM_198862.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028160.mm.1 |
NM_198862.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028160.mm.1 |
TC1100003069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028161.mm.1 |
NM_198862.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028161.mm.1 |
NM_198862.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028161.mm.1 |
TC1100003069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028159.mm.1 |
NM_198862.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028159.mm.1 |
NM_198862.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028159.mm.1 |
TC1100003069.mm.1 |
MTA TC |