Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164659.1 | NP_001158131.1 | 7 - 8 | 1263 | 72 | |
GenBank mRNA | AK048832.1 | - | 7 - 8 | 1067 | 72 | |
AK079552.1 | - | 7 - 8 | 1264 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |