Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgsm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197943.2 | NP_922934.2 | 6 - 7 | 870 | 68 | |
GenBank mRNA | AK122273.1 | - | 6 - 7 | 792 | 68 | |
BC058414.1 | - | 6 - 7 | 861 | 6822416 | 68 | |
BC060163.1 | - | 6 - 7 | 861 | 6822416 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |