Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200018762.mm.1 |
NM_001243712.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018762.mm.1 |
NM_001243712.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018762.mm.1 |
NM_008395.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018762.mm.1 |
NM_008395.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018762.mm.1 |
TC0200002359.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018772.mm.1 |
NM_001243712.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018772.mm.1 |
NM_001243712.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018772.mm.1 |
TC0200002359.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018770.mm.1 |
NM_001243712.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018770.mm.1 |
NM_001243712.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018770.mm.1 |
NM_008395.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018770.mm.1 |
NM_008395.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018770.mm.1 |
TC0200002359.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018774.mm.1 |
NM_001243712.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018774.mm.1 |
NM_001243712.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018774.mm.1 |
NM_008395.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018774.mm.1 |
NM_008395.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018774.mm.1 |
TC0200002359.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018766.mm.1 |
NM_001243712.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018766.mm.1 |
NM_001243712.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018766.mm.1 |
NM_008395.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018766.mm.1 |
NM_008395.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018766.mm.1 |
TC0200002359.mm.1 |
MTA TC |