Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008489.2 | NP_032515.2 | 5 - 6 | 765 | 72 | |
GenBank mRNA | AK143562.1 | - | 5 - 6 | 766 | 72 | |
AK158105.1 | - | 5 - 6 | 648 | 72 | ||
X99347.1 | - | 5 - 6 | 715 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |