Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800010250.mm.1 |
NM_001286260.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010250.mm.1 |
NM_018823.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010250.mm.1 |
NM_018823.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010250.mm.1 |
NM_133957.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010250.mm.1 |
NM_133957.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010250.mm.1 |
TC0800001269.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010251.mm.1 |
NM_001286260.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010251.mm.1 |
NM_018823.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010251.mm.1 |
NM_018823.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010251.mm.1 |
NM_133957.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010251.mm.1 |
NM_133957.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010251.mm.1 |
TC0800001269.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010254.mm.1 |
NM_001286260.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010254.mm.1 |
NM_018823.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010254.mm.1 |
NM_018823.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010254.mm.1 |
NM_133957.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010254.mm.1 |
NM_133957.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010254.mm.1 |
TC0800001269.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010252.mm.1 |
NM_001286260.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010252.mm.1 |
NM_018823.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010252.mm.1 |
NM_018823.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010252.mm.1 |
NM_133957.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010252.mm.1 |
NM_133957.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010252.mm.1 |
TC0800001269.mm.1 |
MTA TC |