Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172410.2 | NP_765998.1 | 16 - 17 | 1832 | 81 | |
GenBank mRNA | AK010450.1 | - | 15 - 16 | 1783 | 81 | |
AK028864.1 | - | 16 - 17 | 1833 | 81 | ||
AK045380.1 | - | 1691 | 81 | |||
AK129055.1 | - | 14 - 15 | 2176 | 81 | ||
AK167105.1 | - | 15 - 16 | 1783 | 81 | ||
AK169194.1 | - | 13 - 14 | 1553 | 81 | ||
AK169693.1 | - | 15 - 16 | 1802 | 81 | ||
BC023140.1 | - | 15 - 16 | 1778 | 3991335 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |