Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfix | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081981.2 | NP_001075450.1 | 6 - 7 | 1941 | 63 | |
NM_001081982.2 | NP_001075451.1 | 6 - 7 | 1266 | 63 | ||
GenBank mRNA | BC003766.1 | - | 6 - 7 | 988 | 3491917 | 63 |
BC090626.1 | - | 6 - 7 | 968 | 63 | ||
S81451.1 | - | 5 - 6 | 952 | 63 | ||
Y07688.1 | - | 6 - 7 | 1268 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |