Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163262.1 | NP_001156734.1 | 12 - 13 | 1568 | 85 | |
NM_028941.1 | NP_083217.1 | 12 - 13 | 1391 | 85 | ||
GenBank mRNA | AK016712.1 | - | 12 - 13 | 1392 | 85 | |
AK173234.1 | - | 6 - 7 | 3818 | 85 | ||
BC028834.1 | - | 10 - 11 | 1060 | 85 | ||
BC158118.1 | - | 13 - 14 | 1445 | 9088191 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |