Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277188.1 | NP_001264117.1 | 20 - 21 | 2412 | 90 | |
NR_102339.1 | 19 - 20 | 2369 | 90 | |||
GenBank mRNA | BC065114.1 | - | 19 - 20 | 2369 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |