Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Best3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007583.1 | NP_001007584.1 | 4 - 5 | 1000 | 86 | |
GenBank mRNA | AY450426.1 | - | 4 - 5 | 1000 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |