Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100019202.mm.1 |
NM_001164636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019202.mm.1 |
NM_001164636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019202.mm.1 |
TC1100002004.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100019203.mm.1 |
NM_001164636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019203.mm.1 |
NM_001164636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019203.mm.1 |
TC1100002004.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100019199.mm.1 |
NM_001164636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019199.mm.1 |
NM_001164636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019199.mm.1 |
TC1100002004.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100019209.mm.1 |
NM_001164636.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019209.mm.1 |
NM_001164636.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019209.mm.1 |
NM_177394.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019209.mm.1 |
NM_177394.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019209.mm.1 |
TC1100002004.mm.1 |
MTA TC |