Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009660.3 | NP_033790.3 | 3 - 4 | 442 | 64 | |
GenBank mRNA | AK142371.1 | - | 3 - 4 | 443 | 64 | |
BC056625.1 | - | 3 - 4 | 429 | 5005087 | 64 | |
BC081546.1 | - | 3 - 4 | 428 | 3671948 | 64 | |
CT010263.1 | - | 3 - 4 | 416 | 64 | ||
L34570.1 | - | 3 - 4 | 426 | 64 | ||
U04331.1 | - | 3 - 4 | 416 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |