Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178367.4 | NP_848144.3 | 1 - 2 | 316 | 72 | |
GenBank mRNA | AK035177.1 | - | 1 - 2 | 316 | 72 | |
AK087592.1 | - | 1 - 2 | 322 | 72 | ||
BC052172.1 | - | 1 - 2 | 310 | 6515901 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |