Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007863.2 | NP_031889.2 | 6 - 7 | 636 | 90 | |
GenBank mRNA | AK032270.1 | - | 5 - 6 | 1910 | 90 | |
AK163205.1 | - | 6 - 7 | 619 | 90 | ||
AY236514.1 | - | 5 - 6 | 470 | 90 | ||
BC052003.1 | - | 883 | 90 | |||
BC062642.1 | - | 6 - 7 | 604 | 5718318 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |