Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp11l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146008.2 | NP_666120.1 | 4 - 5 | 118 | ||
GenBank mRNA | AK040850.1 | - | 4 - 5 | 118 | ||
AK146035.1 | - | 4 - 5 | 118 | |||
BC008239.1 | - | 3 - 4 | 431 | 118 | ||
BC029663.1 | - | 4 - 5 | 5364434 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |