Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dvl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007888.3 | NP_031914.3 | 2 - 3 | 468 | 103 | |
GenBank mRNA | AK050614.1 | - | 2 - 3 | 272 | 103 | |
AK144001.1 | - | 2 - 3 | 498 | 103 | ||
AK146822.1 | - | 2 - 3 | 468 | 103 | ||
AK159895.1 | - | 2 - 3 | 301 | 103 | ||
AK168376.1 | - | 2 - 3 | 376 | 103 | ||
BC021015.1 | - | 2 - 3 | 483 | 103 | ||
BC053050.1 | - | 2 - 3 | 382 | 6402000 | 103 | |
BC092396.1 | - | 475 | 30363483 | 103 | ||
U24160.1 | - | 2 - 3 | 480 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |