Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081332.1 | NP_001074801.1 | 2 - 3 | 1617 | 103 | |
GenBank mRNA | BC151197.1 | - | 2 - 3 | 1058 | 9088085 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |