Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025894.2 | NP_080170.1 | 6 - 7 | 708 | 119 | |
GenBank mRNA | AK005114.1 | - | 6 - 7 | 742 | 118 | |
AK007680.1 | - | 6 - 7 | 682 | 119 | ||
AK008646.1 | - | 6 - 7 | 709 | 119 | ||
AK009969.1 | - | 6 - 7 | 711 | 119 | ||
AK151242.1 | - | 6 - 7 | 709 | 119 | ||
AK152240.1 | - | 6 - 7 | 740 | 119 | ||
AK160283.1 | - | 6 - 7 | 730 | 119 | ||
AK162779.1 | - | 709 | 119 | |||
AK167236.1 | - | 6 - 7 | 716 | 119 | ||
AK186271.1 | - | 3 - 4 | 275 | 119 | ||
BC004694.1 | - | 6 - 7 | 726 | 3498952 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |