Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zswim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172503.3 | NP_766091.2 | 12 - 13 | 2406 | 134 | |
GenBank mRNA | AK052168.1 | - | 12 - 13 | 2420 | 134 | |
AK053663.1 | - | 12 - 13 | 2420 | 134 | ||
AK131119.1 | - | 7 - 8 | 1264 | 134 | ||
AK150221.1 | - | 12 - 13 | 2421 | 134 | ||
BC016518.1 | - | 2 - 3 | 225 | 134 | ||
BC058080.1 | - | 12 - 13 | 2406 | 6822376 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |